missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533711
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Dog, Panda, Pig (100%) Bovine, Bat, Rabbit, Zebrafish (95%) Mouse, Rat (85%).
GPR4 Polyclonal antibody specifically detects GPR4 in Canine, Human, Porcine samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR4 | |
Polyclonal | |
Unconjugated | |
GPR4 | |
G protein-coupled receptor 4; GPR4; Gpr6c; G-protein coupled receptor 19; G-protein coupled receptor 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100144489, 2828, 484440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P46093, P50132 | |
GPR4 | |
Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human GPR4. | |
50 μg | |
Primary | |
Canine, Human, Porcine | |
Antibody | |
IgG |