missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533719
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (91%) Dog, Pig (88%) Rabbit (85%) Bovine (82%).
CRTH2 Polyclonal antibody specifically detects CRTH2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
CRTH2 | |
Polyclonal | |
Unconjugated | |
PTGDR2 | |
CD294; chemoattractant receptor homologous molecule expressed on T helper type 2 cells; chemoattractant receptor-homologous molecule expressed on TH2 cells; CRTH2; DL1R; DP2; G protein-coupled receptor 44; Gpr44; G-protein coupled receptor 44; Grp45; MGC130436; PGD2 receptor; prostaglandin D2 receptor 2; prostaglandin D2 receptor CRTH2; Ptgdr2; putative G-protein coupled receptor 44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9Y5Y4 | |
PTGDR2 | |
Synthetic 33 amino acid peptide from N-terminal extracellular domain of human GPR44 / CRTH2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |