Learn More
Granzyme B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547214
Additional Details : Weight : 0.00010kg
Description
In sandwich ELISAs, less than 1% cross-reactivity with recombinant human (rh) rhGranzyme A, rhGranzyme B, recombinant mouse (rm) Granzyme D, rmGranzyme G, and rhGranzyme H is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Granzyme B Polyclonal antibody specifically detects Granzyme B in Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot.Specifications
Granzyme B | |
Polyclonal | |
Unconjugated | |
GZMB | |
AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P04187 | |
GZMB | |
Mouse myeloma cell line NS0-derived recombinant mouse Granzyme B Lys17-Ser247 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |