Learn More
GRASP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532105
Additional Details : Weight : 0.01000kg
Descrizione
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GRASP1 Polyclonal antibody specifically detects GRASP1 in Human samples. It is validated for Western Blot.Specifica
GRASP1 | |
Polyclonal | |
Unconjugated | |
GRIPAP1 | |
30kDa C-terminus form; AI854681; DXImx47e; Grasp1; GRASP-1; GRASP-1 C-terminal chain; GRIP1; GRIP-1; GRIP1 associated protein 1; GRIP1-associated protein 1; Gripap1; GRIP-associated protein 1; HCMV-interacting protein; Kiaa1167; mKIAA1167; MPMGp800B12492Q3; Sfc10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q4V328 | |
GRIPAP1 | |
Recombinant protein fragment corresponding to a region within amino acids 343 and 560 of Human GRASP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |