missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRSF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541752
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SCRRTGAACL PFYSAASYPA LRASLLPQSL AAAAAVPTRS YSQESKTTYL Sequence homology: Cow: 85%; Human: 100%; Mouse: 85%; Pig: 77%; Rat: 77%
GRSF1 Polyclonal antibody specifically detects GRSF1 in Human samples. It is validated for Western Blot.Specifications
GRSF1 | |
Polyclonal | |
Unconjugated | |
GRSF1 | |
BB232551; C80280; D5Wsu31e; G-rich RNA sequence binding factor 1; G-rich sequence factor 1; Grsf1; GRSF-1 | |
Rabbit | |
Protein A | |
RUO | |
2926 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q12849 | |
GRSF1 | |
Synthetic peptide directed towards the N-terminal of human GRSF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |