Learn More
GTF2H3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124392
Additional Details : Weight : 0.01000kg
Description
PA1-24392 detects GTF2H3 in Human samples. PA1-24392 has been successfully used in Western Blot procedures. PA1-24392 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human GTF2H3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.Specifications
GTF2H3 | |
Polyclonal | |
Unconjugated | |
Q13889 | |
GTF2H3 | |
Synthetic peptide region of human GTF2H3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
GTF2H3 | |
BTF2, TFB4, TFIIH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2967 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |