missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Syrian Hamster IgG (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen A18892
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Syrian Hamster IgG (H+L) Polyclonal antibody specifically detects Syrian Hamster IgG (H+L) in Hamster samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Syrian Hamster IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
Affinity chromatography | |
RUO | |
Hamster | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 0.1mM ZnCl2, 1% BSA, 30mM triethanolamine, 5mM MgCl2 and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |