Learn More
Kininogen Light Chain Monoclonal Antibody (C11C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110831
Additional Details : Weight : 0.01000kg
Description
MA1-10831 detects High MW Kininogen, light chain, from human samples. MA1-10831 has been successfully used in ELISA and Western blot applications. The MA1-10831 immunogen is purified high molecular weight kininogen.
Kininogen Light Chain Monoclonal antibody specifically detects Kininogen Light Chain in Human samples. It is validated for ELISA, Western Blot.Specifications
Kininogen Light Chain | |
Monoclonal | |
0.9 mg/mL | |
PBS with no preservative; pH 7.4 | |
P01042 | |
KNG1 | |
Purified high molecular weight kininogen. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
C11C1 | |
Unconjugated | |
KNG1 | |
alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; bradykinin; Fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; KNG; KNG1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; williams-Fitzgerald-Flaujeac factor | |
Mouse | |
Protein G | |
RUO | |
3827 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |