missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DRA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529814
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HLA-DRA Polyclonal antibody specifically detects HLA-DRA in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HLA-DRA | |
Polyclonal | |
Unconjugated | |
HLA-DRA | |
histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; HLA-DRA; HLA-DRA1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC class II antigen DRA; MLRW | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P01903 | |
HLA-DRA | |
Recombinant fragment corresponding to a region within amino acids 49 and 254 of Human HLA-DRA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |