missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMBS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537366
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 39 kDa. Purity is >95% by SDS-PAGE.
HMBS Polyclonal antibody specifically detects HMBS in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
HMBS | |
Polyclonal | |
Unconjugated | |
Hmbs | |
alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15288, 3145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P08397, P22907 | |
Hmbs | |
Recombinant full length human HMBS | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |