missing translation for 'onlineSavingsMsg'
Läs mer

HNF4A Mouse anti-Human, Rat, Clone: H1415, Invitrogen™

Produktkod. 10732144
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
Varje
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
10732144 100 μg Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10732144 Leverantör Invitrogen Leverantörsnummer 417700

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

This antibody specifically recognizes human HNF4 alpha 1, 2, 4, 5, 7 ,8 and cross reacts with mouse and rat HNF4 alpha 1, 2, 4, 5, 7 ,8.

The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal domit non-insulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen HNF4A
Användningsområden ChIP Assay, ELISA, Gel Shift, Immunohistochemistry, Immunoprecipitation, Western Blot
Klassificering Monoclonal
Klona H1415
Koncentration 1 mg/mL
Konjugera Unconjugated
Gen HNF4A
Genaccessionsnr. P41235, P22449
Gene Alias HNF-4, HNF-4-alpha, HNF4, HNF4A, Hepatocyte nuclear factor 4-alpha, NR2A1, TCF14, Transcription factor 14, Transcription factor HNF-4
Gensymboler HNF4A
Värd art Mouse
Immunogen Baculovirus-expressed recombit human HNF4-alpha (393-461 aa)
Reningsmetod Ammonium sulfate precipitation
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 25735, 3172
Målarter Human, Rat
Innehåll och lagring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG2a
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.