missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP M Monoclonal Antibody (1D8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134981
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
hnRNP M Monoclonal antibody specifically detects hnRNP M in Bovine, Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
hnRNP M | |
Monoclonal | |
Conc. not determined | |
Ascites with 0.1% sodium azide | |
P52272, Q62826, Q9D0E1 | |
HNRNPM | |
M19 fusion protein containing full-length human protein. | |
RUO | |
100356502, 100514153, 116655, 4670, 515290, 76936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1D8 | |
Unconjugated | |
HNRNPM | |
2610023M21Rik; AA409009; CEA receptor; CEAR; heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNPM; HNRNPM4; Hnrpm; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 | |
Mouse | |
200 μL | |
Primary | |
Bovine, Human, Mouse, Porcine, Rabbit, Rat | |
Antibody | |
IgG1 |