missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540375
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MSSYFVNSFS GRYPNGPDYQ LLNYGSGSSL SGSYRDPAAM HTGSYGYNYN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
HOXB5 Polyclonal antibody specifically detects HOXB5 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
HOXB5 | |
Polyclonal | |
Unconjugated | |
HOXB5 | |
AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3215 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P09067 | |
HOXB5 | |
Synthetic peptide directed towards the N-terminal of human HOXB5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |