Learn More
HPD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527980
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (90%), Rat (89%), Pig (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HPD Polyclonal antibody specifically detects HPD in Human samples. It is validated for Western Blot.Specifications
HPD | |
Polyclonal | |
Unconjugated | |
Hpd | |
4H GLOD3; 4-H HPPDASE; 4HPPD; 4-HPPD; 4-hydroxyphenylpyruvate dioxygenase; 4-hydroxyphenylpyruvic acid dioxygenase; 4-hydroxyphenylpyruvic acid oxidase; f Alloantigen; F liver antigen; f protein; Fla; Flp; GLOD3; glyoxalase domain containing 3; HPD; Hppd; HPPDASE; Laf; PPD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3242 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P32754 | |
Hpd | |
Recombinant fragment corresponding to a region within amino acids 49 and 323 of Human HPD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |