Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA174405
2343.45 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Goat anti Human Kappa Light Chain antibody recognizes human kappa light chains. This antibody has been cross absorbed against pooled human myeloma proteins with lambda light chains.
Human Kappa Light Chain Polyclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen).Specifications
Human Kappa Light Chain | |
Polyclonal | |
FITC | |
Iglc | |
Human kappa light chains. | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
1 mg/mL | |
PBS with <0.1% sodium azide; pH 7.4 | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |