Learn More
Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, TRITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A18792
Additional Details : Weight : 0.01000kg
Description
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. 1 year from date of receipt. Prepare working dilution prior to use and then discard. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin human serum immunoglobulins, light chains on all human immunoglobulins, serum proteins from bovine, mouse or rabbit.
Human IgA Cross-Adsorbed Polyclonal antibody specifically detects Human IgA Cross-Adsorbed in Human samples. It is validated for Immunocytochemistry.Specifications
Human IgA Cross-Adsorbed | |
Polyclonal | |
TRITC | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Lyophilized |
Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Human IgA | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |