Learn More
Goat anti-Human IgG (H+L) Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A18807
584.02 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Human IgG (H+L) Polyclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Western Blot.Specifications
Human IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.6 | |
Gamma Immunoglobins Heavy and Light chains | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |