missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG3 Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA183242
1480.07 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Mouse anti Human IgG3 antibody, clone 5G12 recognizes human IgG3, hinge-region with no allotype restriction.
Human IgG3 Monoclonal antibody specifically detects Human IgG3 in Human samples. It is validated for ELISA.Specifications
Human IgG3 | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
IgG3 purified from human plasma. | |
500 μg | |
Secondary | |
-20°C | |
IgG1 |
ELISA | |
5G12 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |