missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129968
Additional Details : Weight : 0.01000kg
Description
PA1-29968 detects ICAM1 from human, rhesus and cynomolgus monkey, baboon, an chimpanzee samples.
ICAM-1 Polyclonal antibody specifically detects ICAM-1 in Baboon, Chimpanzee, Human, Non-human Primate samples. It is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
ICAM-1 | |
Polyclonal | |
Biotin | |
ICAM1 | |
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
Rabbit | |
IgG fraction | |
RUO | |
101021731, 3383, 450196 | |
4° C | |
Liquid |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P05362, Q28806 | |
ICAM1 | |
Full length protein (Human) | |
250 μL | |
Primary | |
Baboon, Chimpanzee, Human, Non-human Primate | |
Antibody | |
IgG |