missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547617
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ID1 Polyclonal antibody specifically detects ID1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
ID1 | |
Polyclonal | |
Unconjugated | |
ID1 | |
AI323524; bHLHb24; class B basic helix-loop-helix protein 24; D2Wsu140e; dJ857M17.1.2; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; ID; ID1; Id-1; ID125A; Idb1; inhibitor of differentiation 1; Inhibitor of DNA binding 1; Inhibitor of DNA binding 1 helix-loop-helix protein (splice varaiation); inhibitor of DNA binding 1, dominant negative helix-loop-helix protein; Inhibitor of DNA binding 1, helix-loop-helix protein (splice variation); inhibitor of DNA binding 1, HLH protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
15901, 3397 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P20067, P41134 | |
ID1 | |
E. coli-derived recombinant mouse ID1 Met1-Glu135 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |