missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 Sulfatase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519035
Additional Details : Weight : 0.01000kg
Description
Iduronate 2 Sulfatase Polyclonal antibody specifically detects Iduronate 2 Sulfatase in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Iduronate 2 Sulfatase | |
Polyclonal | |
Unconjugated | |
Ids | |
alpha-L-iduronate sulfate sulfatase; AW214631; Ids; Iduronate 2-sulfatase; iduronate 2-sulfatase (Hunter syndrome); iduronate 2-sulfatase 14 kDa chain; Iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; idursulfase; MPS2; SIDS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3423 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P22304 | |
Ids | |
Synthetic peptide sequence (KHFRFRDLEEDP) corresponding to the internal amino acids of IDS | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |