Learn More
IGF2BP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541820
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PDEQIAQGPE NGRRGGFGSR GQPRQGSPVA AGAPAKQQQV DIPLRLLVPT Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%
IGF2BP1 Polyclonal antibody specifically detects IGF2BP1 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
IGF2BP1 | |
Polyclonal | |
Unconjugated | |
IGF2BP1 | |
AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10642, 303477 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8CGX0, Q9NZI8 | |
IGF2BP1 | |
Synthetic peptide directed towards the N-terminal of human IGF2BP1 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |