missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Isotype Control (ZX3), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen SA112182
Additional Details : Weight : 0.01000kg
Description
SA1-12182 has been successfully used in FACS procedures to assess non-specific binding of mouse monoclonal antibodies in human samples.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 | |
ZX3 | |
Unconjugated | |
PBS with 0.08% sodium azide | |
Mouse | |
Protein A/G | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
1.0 mg/mL | |
Assay dependent | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
White blood cells. | |
100 μg | |
Isotype Control (Primary) | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |