Läs mer
anti-IgG1 λ, Hamster, PE-Cy™7, Clone: G235-2356, BD
Brand: BD Biosciences 557798
Beskrivning
The immunogen used to produce the G235-2356 hybridoma was the hapten trinitrophenol conjugated to a protein carrier. The G235-2356 antibody was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, <special id='310'/>1Immunogen: Trinitrophenol-KLH
Flow Cytometry, Isotype Control
Specifikationer
| Hamster IgG1, l1 | |
| G235-2356 | |
| PE-Cyanine7 | |
| 0.1 mg | |
| Immunologi |
| Monoclonal | |
| 0.2mg/mL | |
| Affinity Purified | |
| RUO | |
| Primary |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.