Läs mer
anti-IgG2b,κ, Clone: R35-38, BD
Brand: BD Biosciences 562951
Beskrivning
Purified rat IgG2bκ isotype control. The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Host Species: Rat
Clone: R35-38
Isotype: IgG2b, κ
Flow Cytometry, Isotype Control
Specifikationer
| Flow Cytometry | |
| R35-38 | |
| Brilliant Violet 510 | |
| Råtta | |
| 50μ g | |
| Isotype Control (Primary) | |
| IgG2b κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.