Learn More
IL12RB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521479
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Dog (80%), Pig (80%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL12RB2 Polyclonal antibody specifically detects IL12RB2 in Human samples. It is validated for Western Blot.Specifications
IL12RB2 | |
Polyclonal | |
Unconjugated | |
IL12RB2 | |
A930027I18Rik; Ifnm; IL-12 receptor subunit beta-2; IL-12R subunit beta-2; Il12rb2; IL-12RB2; IL-12R-beta-2; interferon response to microorganisms; interleukin 12 receptor subunit beta 2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; interleukin-12 receptor subunit beta-2; RP11-102M16.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q99665 | |
IL12RB2 | |
Recombinant fragment corresponding to a region within amino acids 323 and 695 of IL-12 Receptor beta2 (Uniprot ID#Q99665) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |