missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22 Receptor alpha Rat anti-Mouse, Clone: 496514, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524205
This item is not returnable.
View return policy
Description
In direct ELISAs, 25% cross-reactivity with recombit human (rh) IL-22 R alpha 1 is observed and no cross-reactivity with rhIL-20 R alpha, recombit mouse (rm) IL-20 R alpha, rhIL-22BP, or rmIL-22BP is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
Specifications
| IL-22 Receptor alpha | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q80XZ4 | |
| IL22RA1 | |
| Mouse myeloma cell line NS0-derived recombit mouse IL-22 R alpha 1 Thr18-Ala228 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 496514 | |
| Unconjugated | |
| Il22ra1 | |
| CRF2-9, IL22R, IL22R1, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, cytokine receptor class-II member 9, cytokine receptor family 2 member 9, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 230828 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction