missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL24 Monoclonal Antibody (303308), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524013
Additional Details : Weight : 0.00010kg
Description
No cross-reactivity with recombinant human IL-24 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
IL24 Monoclonal antibody specifically detects IL24 in Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
IL24 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q925S4 | |
IL24 | |
E. coli-derived recombinant mouse IL-24 Arg27-His181 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
303308 | |
Unconjugated | |
IL24 | |
C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP | |
Rat | |
Protein A/G | |
RUO | |
93672 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |