Learn More
IL-5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547458
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, 50% cross-reactivity with recombinant canine IL-5 and recombinant feline IL-5 is observed and 30% cross-reactivity with recombinant human IL-5, recombinant mouse IL-5, and recombinant rat IL-5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-5 Polyclonal antibody specifically detects IL-5 in Porcine samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
IL-5 | |
Polyclonal | |
Unconjugated | |
Il5 | |
B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
397409 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9MYM5 | |
Il5 | |
Mouse myeloma cell line NS0-derived recombinant porcine IL-5 Ile20-Ser134 | |
100 μg | |
Primary | |
Porcine | |
Antibody | |
IgG |