Learn More
IL5RA Monoclonal Antibody (26815), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523581
2540.12 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) IL-5 R beta, rhIL-9 R, rhIL-4 R, rmIL-5 R alpha, or rhIL-13 R alpha 1.
IL5RA Monoclonal antibody specifically detects IL5RA in Human samples. It is validated for Flow Cytometry.Specifications
IL5RA | |
Monoclonal | |
APC | |
IL5RA | |
CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha | |
Mouse | |
Protein A/G | |
RUO | |
3568 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
26815 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q01344 | |
IL5RA | |
Sf 21-derived recombinant human IL-5R alpha/CD125 Asp21-Arg335 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |