missing translation for 'onlineSavingsMsg'
Learn More

IL-8 (CXCL8), Biotin, clone: 790A28G2, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen AHC0789

Additional Details : Weight : 0.50000kg

Product Code. 10755143

  • 3030.00 SEK / Each


EU Stock 1
Add to basket

For Research Use Only. All usage must comply with product instructions.


Description

Description

This product is recommended as a detection antibody in Sandwich ELISA applications.
Specifications

Specifications

IL-8 (CXCL8)
Monoclonal
0.5mg/mL
PBS with 5mg/mL BSA and 0.05% sodium azide; pH 7.5
P10145
Interleukin-8, Monocyte-derived neutrophil chemotactic factor, T-cell chemotactic factor, Neutrophil-activating protein 1, CXCL8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
Murine
IgG1, κ
100μg
4° C
Primary
3576
ELISA
790A28G2
Biotin
CXCL8
Liquid
CXCL8
Recombinant human IL-8.
Protein A
RUO
Antibody
Monoclonal
Human
Videos