missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9 Monoclonal Antibody (623153), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523679
Additional Details : Weight : 0.00010kg
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry.Specifications
IL-9 | |
Monoclonal | |
APC | |
Il9 | |
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
Mouse | |
Protein A/G | |
RUO | |
3578 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
623153 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P15248 | |
Il9 | |
Sf 21-derived recombinant human IL-9 Gln19-Ile144 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |