Learn More
IL-9 Recombinant Rabbit Monoclonal Antibody (2H14L3), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701229
Additional Details : Weight : 0.01000kg
Description
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
IL-9 Recombinant Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Western Blot.Specifications
IL-9 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
proprietary | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
2H14L3 | |
Unconjugated | |
P15248 | |
Rabbit | |
Protein A | |
RUO | |
3578 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |