Learn More
IL-12 p35 Monoclonal Antibody (27537), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523559
Additional Details : Weight : 0.00010kg
Description
This antibody detects the p35 subunit either as part of a p40/p35 heterodimer or as a free subunit after reduction of the heterodimer. This antibody does not recognize IL-12 p40 homodimers but shows strong cross-reactivity with the p35 subunits from porcine and mouse systems.
IL-12 p35 Monoclonal antibody specifically detects IL-12 p35 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
IL-12 p35 | |
Monoclonal | |
PE | |
IL12A | |
CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
Mouse | |
Protein A/G | |
RUO | |
16159, 3592 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
27537 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P29459, P43431 | |
IL12A | |
Sf 21-derived recombinant human IL-12/IL-35 p35 Arg23-Ser219 | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |