missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519131
Additional Details : Weight : 0.01000kg
Description
IL-18 Polyclonal antibody specifically detects IL-18 in Human samples. It is validated for Western Blot.Specifications
IL-18 | |
Polyclonal | |
Unconjugated | |
IL18 | |
cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3606 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q14116 | |
IL18 | |
Synthetic peptide sequence (NPPDNIKDTKSDI) corresponding to the internal amino acids of IL18 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |