missing translation for 'onlineSavingsMsg'
Läs mer

IL27, Human, IP-WB Antibody Pair (PW3), Abnova™

Produktkod. 16183904
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16183904 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16183904 Leverantör Abnova Leverantörsnummer H00246778PW3.1Set
 Begränsad artikel
Beräknad leverans: 09-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Innehåller

Antibody pair for IP: rabbit polyclonal anti-IL27 (300μL), Antibody pair for WB: mouse purified polyclonal anti-IL27 (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifikationer

Antigen IL27
Användningsområden Immunoprecipitation, Western Blot
Klassificering Monoclonal/Polyclonal
Konjugera Unconjugated
Beskrivning This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gen IL27
Gene Alias IL-27/IL-27A/IL27p28/IL30/MGC71873/p28
Gensymboler IL27
Värd art Mouse, Rabbit
Kvantitet 1 Set
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 246778
Inkluderar Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-IL27 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-IL27 (50 ug)
Målarter Human
Innehåll och lagring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.