missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA123094
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H5 Hemagglutinin HA1 Recombinant Protein(GTX48233-PRO). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Avian Influenza A HA Polyclonal antibody specifically detects Avian Influenza A HA in Virus samples. It is validated for ELISA, Western Blot.Specifications
Avian Influenza A HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin | |
Synthetic peptide corresponding to 15 amino acids at the N-terminus of the hemagglutinin protein. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |