Läs mer
Integrin β7 Rat, Brilliant Violet 421, Clone: FIB504, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 564283
Beskrivning
FIB504 reacts with mouse integrin β 7 subunit (130 kD) but also cross reacts with human integrin β 7. Integrin β 7 associates with α 4 (CD49d) expressed on subsets of lymphocytes and thymus. It also associates with α IEL (CD103) expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Integrin β 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifikationer
| Integrin β7 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| Mouse T lymphoma line TK1 | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| FIB504 | |
| Itgb7; Integrin β7; Integrin beta 7; ITB7; Ly-69; Ly69 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG2b κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.