missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA4 Rat anti-Mouse, PE, Clone: 265329, Invitrogen™
Rat Monoclonal Antibody
Brand: Thermo Scientific MA523604
Additional Details : Weight : 0.00010kg
Description
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.Specifications
ITGA4 | |
Monoclonal | |
PE | |
Itga4 | |
269C wild type, CD49 antigen-like family member D, VLA-4 subunit alpha, antigen CD49D, alpha-4 subunit of VLA-4 receptor, ITGA4, ITGA-IV | |
Rat | |
Protein A/G | |
RUO | |
16401 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
265329 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q00651 | |
ITGA4 | |
Mouse myeloma cell line NS0-derived recombit mouse Integrin alpha 4/CD49d Tyr34-Phe976 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2a |