missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human Lambda Light Chain Secondary Antibody, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen PA516604
Additional Details : Weight : 0.01000kg
Description
PA5-16604 targets Lambda in IF applications and shows reactivity with Human samples. The PA5-16604 immunogen is lambda light chains isolated from human urine.
Specifications
Human Lambda Light Chain | |
Polyclonal | |
TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4 | |
Rabbit | |
Purified | |
RUO | |
Human | |
Liquid |
Immunofluorescence | |
FITC | |
Iglc | |
Lambda light chains isolated from human urine | |
2 mL | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |