Learn More
Lamin A/C Monoclonal Antibody (Jol3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA15820
Additional Details : Weight : 0.01000kg
Description
MA1-5820 reacts with human dermal fibroblasts in immunocytochemistry. Other cell/tissue types have not been tested in this application. It reacts with human, mouse and rat samples in Western blot. This antibody recognizes the common tail domain of Lamin A+C, epitope aa 464-572.
Lamin A/C Monoclonal antibody specifically detects Lamin A/C in Human, Mouse, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.Specifications
Lamin A/C | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.1% sodium azide | |
P02545, P48678, P48679 | |
LMNA | |
Recombinant bacterially expressed full length protein | |
RUO | |
16905, 4000, 60374 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Western Blot | |
Jol3 | |
Unconjugated | |
LMNA | |
70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
Mouse | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |