missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Laminin A/B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA184171
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PA1-84171 recognizes both A (400 kD) and B (220 kD) chains of laminin. A suggested positive control for immunohistochemical applicationss is human kidney.
Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Laminin anchors cells to the basement membrane through interactions with integrins at the cell surface and collagens in the basement membrane.
Specifikationer
| Laminin A/B | |
| Polyclonal | |
| Unconjugated | |
| LAMA1 | |
| AA408497; alternatively spliced laminin gamma2 chain; AW211941; B2T; basement membrane adhesion protein; BM600; BM600-100kDa; C77966; C80098; C81607; Cell-scattering factor 140 kDa subunit; CLM; CSF; CSF 140 kDa subunit; D130003D08Rik; EBR2; EBR2A; Epiligrin subunit gamma; extra-cellular matrix protein; gamma 1 (formerly LAMB2); hypothetical protein; kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; LAMA; Lama1; Lama-1; LAMB1; Lamb-1; Lamb1-1; LAMB2; Lamb-2; LAMB2T; LAMC1; LAMC2; laminin 5 gamma 2 subunit; laminin A chain; laminin alpha 1; laminin alpha 1 chain; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B1s chain; Laminin B2 chain; laminin B2t chain; laminin beta 2 subunit chain; laminin beta 2-like chain; laminin beta-1-1 chain; Laminin beta-1-2 chain; laminin chain B3; laminin chain beta 2; laminin gamma 1; laminin S; laminin subunit alpha 1; laminin subunit alpha-1; laminin subunit beta 1; laminin subunit beta 2; Laminin subunit beta-1; laminin subunit beta-1 variant; laminin subunit beta-2; laminin subunit gamma 1; laminin subunit gamma 2; laminin subunit gamma-1; Laminin subunit gamma-2; laminin, alpha 1; laminin, beta 1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit beta; laminin-11 subunit gamma; Laminin-12 subunit beta; laminin-14 subunit beta; Laminin-15 subunit beta; laminin-2 subunit beta; Laminin-2 subunit gamma; laminin-3 subunit alpha; Laminin-3 subunit beta; laminin-3 subunit gamma; Laminin-4 subunit beta; laminin-4 subunit gamma; laminin-5 gamma 2; Laminin-5 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit beta; laminin-7 subunit gamma; Laminin-8 subunit beta; laminin-8 subunit gamma; Laminin-9 subunit beta; laminin-9 subunit gamma; LAMNB2; Lams; Large adhesive scatter factor 140 kDa subunit; LIS5; Nicein subunit gamma; NPHS5; npht; PTBHS; SLAM; S-LAM alpha; S-LAM beta; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit beta; S-laminin subunit gamma | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Avian, Canine, Chicken, Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| Conc. Not Determined | |
| whole serum with 0.09% sodium azide | |
| P02469, P07942, P15800, P19137, P25391, P55268, Q01635, Q61292 | |
| LAMA1, LAMB1, LAMB2 | |
| Purified laminin from Engelbreth-Holm-Swarm (EHS) tumor. | |
| RUO | |
| 100009388, 100357482, 100358679, 102156629, 16772, 16777, 16779, 25473, 284217, 298941, 316758, 373980, 374016, 3912, 3913, 396478, 475882, 476626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering