missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Langerin Mouse anti-Human, APC, Clone: 343828, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523530
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The protein encoded by this gene is expressed only in Langerhans cells which are immature dendritic cells of the epidermis and mucosa. It is localized in the Birbeck granules, organelles present in the cytoplasm of Langerhans cells and consisting of superimposed and zippered membranes. It is a C-type lectin with mannose binding specificity, and it has been proposed that mannose binding by this protein leads to internalization of antigen into Birbeck granules and providing access to a nonclassical antigen-processing pathway.
Specifikationer
| Langerin | |
| Monoclonal | |
| APC | |
| CD207 | |
| CLEC4K, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, CD207 antigen, langerin, Langerhans cell specific c-type lectin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 50489 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 343828 | |
| Buffered saline solution with BSA and 0.09% sodium azide | |
| Q9UJ71 | |
| CD207 | |
| Mouse myeloma cell line NS0-derived recombit human Langerin/CD207 Tyr64-Pro328 | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering