Läs mer
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Beskrivning
This gene encodes a member of the glycosyltransferase superfamily. The encoded protein is a single-pass type II Golgi membrane protein that functions as a fucose-specific glycosyltransferase, adding an N-acetylglucosamine to the fucose residue of a group of signaling receptors involved in regulating cell fate decisions during development. Mutations in this gene have been associated with autosomal recessive spondylocostal dysostosis 3. Alternatively spliced transcript variants that encode different isoforms have been described, however, not all variants have been fully characterized. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against a full-length human LFNG protein. |
| Formulering | PBS with no preservative; pH 7.4 |
| Gen | LFNG |
| Genaccessionsnr. | BC014851 |
| Gene Alias | SCDO3 |
| Gensymboler | LFNG |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.