missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGP2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518783
Additional Details : Weight : 0.01000kg
Description
LGP2 Polyclonal antibody specifically detects LGP2 in Human samples. It is validated for Western Blot.Specifications
LGP2 | |
Polyclonal | |
Unconjugated | |
DHX58 | |
B430001I08Rik; D11LGP2; D11lgp2e; DEXH (Asp-Glu-X-His) box polypeptide 58; DEXH-box helicase 58; DHX58; Lgp2; LPG2; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; Probable ATP-dependent RNA helicase DHX58; protein D11Lgp2; Protein D11Lgp2 homolog; RGD1310093; RIG-I-like receptor 3; RIG-I-like receptor Lgp2; RLR; RLR-3; RNA helicase LGP2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
79132 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96C10 | |
DHX58 | |
Synthetic peptide sequence (DFLQHCAENLSD) corresponding to the C-terminus amino acids of DHX58 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |