Learn More
LHX9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541160
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RNLLRQENGG VDKADGTSLP APPSADSGAL TPPGTATTLT DLTNPTITVV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 100%
LHX9 Polyclonal antibody specifically detects LHX9 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
LHX9 | |
Polyclonal | |
Unconjugated | |
LHX9 | |
3110009O07Rik; BB104635; LH2B; Lhx9; LIM homeobox 9; LIM homeobox 9-like; LIM homeobox protein 9; LIM homeodomain type transcription factor Lhx9; LIM/homeobox protein Lhx9; LIM-homeobox protein 9; LIM-homeodomain type transcription factor 9; si:dkey-121a9.2; zgc:112196 | |
Rabbit | |
Protein A | |
RUO | |
56956 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NQ69 | |
LHX9 | |
Synthetic peptide directed towards the C-terminal of human LHX9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |