Learn More
LIMCH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534745
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H1299, HeLaS3, mouse brain. Predicted reactivity: Mouse (80%), Bovine (86%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LIMCH1 Polyclonal antibody specifically detects LIMCH1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LIMCH1 | |
Polyclonal | |
Unconjugated | |
LIMCH1 | |
3732412D22Rik; Kiaa1102; LIM and calponin homology domains 1; LIM and calponin homology domains-containing protein 1; Limch1; LIMCH1A; LMO7B; mKIAA1102 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22998, 77569 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.53 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q3UH68, Q9UPQ0 | |
LIMCH1 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 734 and 1007 (Q9UPQ0) of LIMCH1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |