Learn More
Lipid A LPS Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA128903
Additional Details : Weight : 0.01000kg
Description
PA1-28903 is cross-reactive with numerous members of the Enterobacteriaceae including Pseudomonas aeruginosa, Klebsiella pneumoniae, E. coli O157, Salmonella enteriditis, Enterobacter aerogenes, E. hermanii, Yersinia enterocolitica and Shigella sonnei.
Lipid A LPS Polyclonal antibody specifically detects Lipid A LPS in Bacteria samples. It is validated for Immunocytochemistry.Specifications
Lipid A LPS | |
Polyclonal | |
Unconjugated | |
Lipid A Lipopolysaccharide | |
Whole cells prep of Lipid A from E. coli O157. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |