missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSP1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519274
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and canine based on sequence homology.
LSP1 Polyclonal antibody specifically detects LSP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
LSP1 | |
Polyclonal | |
Unconjugated | |
LSP1 | |
1 BAC CH230-300K22 (Children's Hospital Oakland Research Institute) complete; 47 kDa actin binding protein; 47 kDa actin-binding protein; 52 kDa phosphoprotein; F-actin binding and cytoskeleton associated protein; leufactin (leukocyte F-actin binding protein); leukocyte specific protein 1; leukocyte-specific protein 1; lsp; LSP1; Lsp-1; lymphocyte specific 1; lymphocyte specific protein 1; lymphocyte-specific antigen WP34; Lymphocyte-specific protein 1; OTTHUMP00000014138; OTTHUMP00000069614; OTTHUMP00000069615; OTTHUMP00000069616; p50; pp52; S37; S37 protein; WP34 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16985, 4046 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P19973, P33241 | |
LSP1 | |
Synthetic peptide sequence (RYKFVATGHGKYEKV) corresponding to the C-terminus amino acids of LSP1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |