Learn More
HSFX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538814
Additional Details : Weight : 0.00900kg
Specifications
HSFX1 | |
Polyclonal | |
Unconjugated | |
HSFX1 | |
LW-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100506164 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UBD0 | |
HSFX1 | |
A synthetic peptide derived from the N-terminal region of human LW-1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |